Broxyquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5,7-Dibromo-8-hydroxyquinoline, 5,7-Dibromo-8-quinolinol

HC-6706-100G HC-6706-25G
COBBHC-6706-100GEA 0 GBP
COBBHC-6706-100G COBBHC-6706-25G
Broxyquinoline
Broxyquinoline
Formula: C₉H₅Br₂NO
MW: 302.95 g/mol
Boiling Pt: 371 °C (1013 hPa)
Density: 2.189 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006785
CAS Number: 521-74-4
EINECS: 208-317-8
Merck Index: 13,01438

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR