5-(4-Bromophenyl)-1H-tetrazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7679-5G OS-7679-25G OS-7679-1G
COBBOS-7679-5GEA 0 GBP
COBBOS-7679-5G COBBOS-7679-25G COBBOS-7679-1G
5-(4-Bromophenyl)-1H-tetrazole
5-(4-Bromophenyl)-1H-tetrazole
Formula: C₇H₅BrN₄
MW: 225.05 g/mol
MDL Number: MFCD00991874
CAS Number: 50907-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR