4-Propylbenzenesulphonyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Propylbenzene-1-sulphonyl chloride

003807-5G 003807-25G 003807-10G
FLUO003807-5GEA 0 GBP
FLUO003807-5G FLUO003807-25G FLUO003807-10G
4-Propylbenzenesulphonyl chloride
4-Propylbenzenesulphonyl chloride
Formula: C₉H₁₁ClO₂S
MW: 218.7 g/mol
Boiling Pt: 113…113.5 °C (0.2 mmHg)
Melting Pt: –4 °C
MDL Number: MFCD00041508
CAS Number: 63014-04-0
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR