4-Methylthiobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Methylbenzothioamide, Thio-p-toluamide

021339-1G 021339-5G
FLUO021339-1GEA 0 GBP
FLUO021339-1G FLUO021339-5G
4-Methylthiobenzamide
4-Methylthiobenzamide
Formula: C₈H₉NS
MDL Number: MFCD00173750
CAS Number: 2362-62-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR