4-Methylbenzamidine hydrochloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Methylphenylamidine hydrochloride, 4-Methylbenzene-1-carboximidamide hydrochloride, 4-Methylbenzimidamide hydrochloride

017242-5G 017242-1G
FLUO017242-5GEA 0 GBP
FLUO017242-5G FLUO017242-1G
4-Methylbenzamidine hydrochloride
4-Methylbenzamidine hydrochloride
Formula: C₈H₁₁ClN₂
MW: 170.64 g/mol
MDL Number: MFCD00040183
CAS Number: 6326-27-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR