4-Methoxypyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OS-7400-5G OS-7400-25G OS-7400-100G
COBBOS-7400-5GEA 0 GBP
COBBOS-7400-5G COBBOS-7400-25G COBBOS-7400-100G
4-Methoxypyridine
4-Methoxypyridine
Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD00674049
CAS Number: 620-08-6
EINECS: 210-624-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR