4-Methoxyindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

IN-0053-5G IN-0053-25G IN-0053-1G
COBBIN-0053-5GEA 0 GBP
COBBIN-0053-5G COBBIN-0053-25G COBBIN-0053-1G
4-Methoxyindole
4-Methoxyindole
Formula: C₉H₉NO
Boiling Pt: 307 °C (1013 hPa)
Melting Pt: 67…68 °C
MDL Number: MFCD00009737
CAS Number: 4837-90-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR