4-Methoxy-3-nitropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6702-5G HC-6702-25G HC-6702-1G
COBBHC-6702-5GEA 0 GBP
COBBHC-6702-5G COBBHC-6702-25G COBBHC-6702-1G
4-Methoxy-3-nitropyridine
4-Methoxy-3-nitropyridine
Formula: C₆H₆N₂O₃
MDL Number: MFCD00209661
CAS Number: 31872-62-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR