Indole-4-carboxylic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
IN-0004-1G
IN-0004-25G
IN-0004-100G
IN-0004-5G
COBBIN-0004-1GEA
0
GBP
COBBIN-0004-1G
COBBIN-0004-25G
COBBIN-0004-100G
COBBIN-0004-5G
Indole-4-carboxylic acid
Indole-4-carboxylic acid
Formula:
C₉H₇NO₂ |
MDL Number:
MFCD00009739 CAS Number: 2124-55-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...