4-Hydroxythiobenzamide ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
H32780.06
H32780.14
H32780.06EA
74.8
GBP
H32780.06
H32780.14
4-Hydroxythiobenzamide ≥98%
4-Hydroxythiobenzamide
Purity: 98%
CAS No.: 25984-63-8
Molecular Formula: C7H7NOS
Formula Weight: 153.20
Melting Point: 181-185°
MDL No.: MFCD04973332
Formula:
C₇H₇NOS Melting Pt: 181…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD04973332 CAS Number: 25984-63-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...