4-Hydroxybenzamide ≥98%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
B24222.06
B24222.14
B24222.06EA
26.8
GBP
B24222.06
B24222.14
4-Hydroxybenzamide ≥98%
4-Hydroxybenzamide
Purity: 98+%
CAS No.: 619-57-8
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Melting Point: 159-162°
Beilstein Reference: 1906921
Formula:
C₇H₇NO₂ Melting Pt: 159…162 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007997 CAS Number: 619-57-8 EINECS: 210-602-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...