4-Fluoroindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Fluoro-1H-indole

IN-0051-5G IN-0051-25G IN-0051-1G
COBBIN-0051-5GEA 0 GBP
COBBIN-0051-5G COBBIN-0051-25G COBBIN-0051-1G
4-Fluoroindole
4-Fluoroindole
Formula: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD00055992
CAS Number: 387-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR