4-Fluorobenzenesulphonamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006184-5G 006184-25G 006184-1G
FLUO006184-5GEA 0 GBP
FLUO006184-5G FLUO006184-25G FLUO006184-1G
4-Fluorobenzenesulphonamide
4-Fluorobenzenesulphonamide
Formula: C₆H₆FNO₂S
MW: 175.18 g/mol
MDL Number: MFCD00025384
CAS Number: 402-46-0
EINECS: 206-946-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR