4-Fluoro-3-nitrobenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

013849-25G 013849-5G 013849-1G
FLUO013849-25GEA 0 GBP
FLUO013849-25G FLUO013849-5G FLUO013849-1G
4-Fluoro-3-nitrobenzaldehyde
4-Fluoro-3-nitrobenzaldehyde
Formula: C₇H₄FNO₃
MW: 169.11 g/mol
MDL Number: MFCD01861388
CAS Number: 42564-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR