4-Fluoro-3-methylbenzaldehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Fluoro-m-tolylaldehyde, 4-Fluoro-3-tolualdehyde, 4-Fluoro-m-tolualdehyde

005785-5G 005785.10G 005785-1G 005785-25G
FLUO005785-5GEA 0 GBP
FLUO005785-5G FLUO005785.10G FLUO005785-1G FLUO005785-25G
4-Fluoro-3-methylbenzaldehyde
4-Fluoro-3-methylbenzaldehyde
Formula: C₈H₇FO
MW: 138.14 g/mol
MDL Number: MFCD01631431
CAS Number: 135427-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR