4-Fluoro-o-toluic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

HC-6860-5G HC-6860-25G
COBBHC-6860-5GEA 0 GBP
COBBHC-6860-5G COBBHC-6860-25G
4-Fluoro-o-toluic acid
4-Fluoro-o-toluic acid
Formula: C₈H₇FO₂
MDL Number: MFCD04115880
CAS Number: 321-21-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR