4-Acetylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-Acetylphenyl)metaboric acid, 4-Acetylbenzeneboronic acid, Dihydroxy(4-acetylphenyl)borane

BB-2187-5G BB-2187-25G BB-2187-100G
COBBBB-2187-5GEA 0 GBP
COBBBB-2187-5G COBBBB-2187-25G COBBBB-2187-100G
4-Acetylphenylboronic acid
4-Acetylphenylboronic acid
Formula: C₈H₉BO₃
MW: 163.97 g/mol
MDL Number: MFCD01074667
CAS Number: 149104-90-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR