4-Chloroindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Chloro-1H-indole

IN-0040-5G IN-0040-25G IN-0040-1G
COBBIN-0040-5GEA 0 GBP
COBBIN-0040-5G COBBIN-0040-25G COBBIN-0040-1G
4-Chloroindole
4-Chloroindole
Formula: C₈H₆ClN
MW: 151.6 g/mol
Boiling Pt: 293
Melting Pt: 84…86 °C
Density: 1.259 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00005665
CAS Number: 25235-85-2
EINECS: 246-747-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR