4-Chloro-3-(trifluoromethyl)benzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Chloro-α,α,α-trifluoro-m-toluic acid

005710-5G 005710-1G
FLUO005710-5GEA 0 GBP
FLUO005710-5G FLUO005710-1G
4-Chloro-3-(trifluoromethyl)benzoic acid
4-Chloro-3-(trifluoromethyl)benzoic acid
Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
MDL Number: MFCD01631468
CAS Number: 1737-36-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR