4-Chloro-2-nitrobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CA-4514-25G CA-4514-100G
COBBCA-4514-25GEA 0 GBP
COBBCA-4514-25G COBBCA-4514-100G
4-Chloro-2-nitrobenzoic acid
4-Chloro-2-nitrobenzoic acid
Formula: C₇H₄ClNO₄
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00007214
CAS Number: 6280-88-2
EINECS: 228-483-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR