4-Bromothiobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021311-1G 021311-2.5G 021311-25G 021311-5G
FLUO021311-1GEA 0 GBP
FLUO021311-1G FLUO021311-2.5G FLUO021311-25G FLUO021311-5G
4-Bromothiobenzamide
4-Bromothiobenzamide
Formula: C₇H₆BrNS
MDL Number: MFCD04114360
CAS Number: 26197-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR