4-Bromophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Bromobenzeneboronic acid, (4-Bromophenyl)boronic acid, Dihydroxy(4-bromophenyl)borane, (4-Bromophenyl)metaboric acid

BB-2629-5G BB-2629-25G BB-2629-100G
COBBBB-2629-5GEA 0 GBP
COBBBB-2629-5G COBBBB-2629-25G COBBBB-2629-100G
4-Bromophenylboronic acid
4-Bromophenylboronic acid
Formula: C₆H₆BBrO₂
MW: 200.83 g/mol
Density: 1,67 g/cm³ (20 °C)
MDL Number: MFCD00002104
CAS Number: 5467-74-3
EINECS: 226-779-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR