4-Bromoisoquinoline
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OS-7657-100G
OS-7657-5G
OS-7657-25G
COBBOS-7657-100GEA
0
GBP
COBBOS-7657-100G
COBBOS-7657-5G
COBBOS-7657-25G
4-Bromoisoquinoline
4-Bromoisoquinoline
Formula:
C₉H₆BrN MW: 208.06 g/mol Boiling Pt: 280 Melting Pt: 42…42.5 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006904 CAS Number: 1532-97-4 EINECS: 216-244-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...