4-Bromoindole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-1H-indole

IN-0043-100G IN-0043-5G IN-0043-25G IN-0043-1G
COBBIN-0043-100GEA 0 GBP
COBBIN-0043-100G COBBIN-0043-5G COBBIN-0043-25G COBBIN-0043-1G
4-Bromoindole
4-Bromoindole
Formula: C₈H₆BrN
MW: 196.05 g/mol
MDL Number: MFCD00671502
CAS Number: 52488-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR