4-Bromo-N,N-dimethylbenzenesulphonamide

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: N,N-Dimethyl 4-bromobenzenesulphonamide

OT-0091-5G OT-0091-1G OT-0091-25G
COBBOT-0091-5GEA 0 GBP
COBBOT-0091-5G COBBOT-0091-1G COBBOT-0091-25G
4-Bromo-N,N-dimethylbenzenesulphonamide
4-Bromo-N,N-dimethylbenzenesulphonamide
Formula: C₈H₁₀BrNO₂S
MW: 264.14 g/mol
MDL Number: MFCD00017834
CAS Number: 707-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR