Methyl 4-bromo-3-nitrobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4538-25G CA-4538-1G CA-4538-5G
COBBCA-4538-25GEA 0 GBP
COBBCA-4538-25G COBBCA-4538-1G COBBCA-4538-5G
Methyl 4-bromo-3-nitrobenzoate
Methyl 4-bromo-3-nitrobenzoate
Formula: C₈H₆BrNO₄
MDL Number: MFCD00553089
CAS Number: 2363-16-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR