4-Bromo-3-nitrobenzoic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

008948-1G 008948-5G
FLUO008948-1GEA 0 GBP
FLUO008948-1G FLUO008948-5G
4-Bromo-3-nitrobenzoic acid
4-Bromo-3-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MDL Number: MFCD00272137
CAS Number: 6319-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR