4-Bromo-3-nitrobenzoic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CA-4449-25G
CA-4449-5G
CA-4449-1G
COBBCA-4449-25GEA
0
GBP
COBBCA-4449-25G
COBBCA-4449-5G
COBBCA-4449-1G
4-Bromo-3-nitrobenzoic acid
4-Bromo-3-nitrobenzoic acid
Formula:
C₇H₄BrNO₄ |
MDL Number:
MFCD00272137 CAS Number: 6319-40-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...