4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HC-2137-5G
HC-2137-1G
COBBHC-2137-5GEA
0
GBP
COBBHC-2137-5G
COBBHC-2137-1G
4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazole
4-Bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazole
Formula:
C₅H₄BrF₃N₂ |
MDL Number:
MFCD00120266 CAS Number: 60061-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...