4-Bromo-3-fluorobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CA-4021-5G CA-4021-25G CA-4021-100G
COBBCA-4021-5GEA 0 GBP
COBBCA-4021-5G COBBCA-4021-25G COBBCA-4021-100G
4-Bromo-3-fluorobenzoic acid
4-Bromo-3-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MW: 219.01 g/mol
MDL Number: MFCD00672932
CAS Number: 153556-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR