4-Bromo-3-fluorobenzaldehyde

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7622-5G OS-7622-25G OS-7622-1G OS-7622-100G
COBBOS-7622-5GEA 0 GBP
COBBOS-7622-5G COBBOS-7622-25G COBBOS-7622-1G COBBOS-7622-100G
4-Bromo-3-fluorobenzaldehyde
4-Bromo-3-fluorobenzaldehyde
Formula: C₇H₄BrFO
MW: 203.01 g/mol
MDL Number: MFCD03095000
CAS Number: 133059-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR