4-Bromo-3,5-dimethylisoxazole
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HI-1305-25G
HI-1305-5G
COBBHI-1305-25GEA
0
GBP
COBBHI-1305-25G
COBBHI-1305-5G
4-Bromo-3,5-dimethylisoxazole
4-Bromo-3,5-dimethylisoxazole
Formula:
C₅H₆BrNO MW: 176.01 g/mol |
MDL Number:
MFCD00068187 CAS Number: 10558-25-5 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...