Methyl-4-bromo-2-nitrobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Methyl 4-bromo-2-nitrobenzoate, 4-Bromobenzoic acid methyl ester

CA-4551-1G CA-4551-25G CA-4551-5G
COBBCA-4551-1GEA 0 GBP
COBBCA-4551-1G COBBCA-4551-25G COBBCA-4551-5G
Methyl-4-bromo-2-nitrobenzoate
Methyl 4-bromo-2-nitrobenzoate
Formula: C₈H₆BrNO₄
MDL Number: MFCD10699645
CAS Number: 158580-57-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR