tert-Butyl 4-bromo-2-nitrobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: tert-Butyl 4-bromo-2-nitrobenzoate, 4-Bromobenzoic acid tert-butyl ester

CA-4285-5G CA-4285-1G CA-4285-25G
COBBCA-4285-5GEA 0 GBP
COBBCA-4285-5G COBBCA-4285-1G COBBCA-4285-25G
tert-Butyl 4-bromo-2-nitrobenzoate
tert-Butyl 4-bromo-2-nitrobenzoate
Formula: C₁₁H₁₂BrNO₄
MW: 302.12 g/mol
MDL Number: MFCD09475812
CAS Number: 890315-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR