4-Bromo-2-nitrobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CA-4017-25G CA-4017-5G CA-4017-100G
COBBCA-4017-25GEA 0 GBP
COBBCA-4017-25G COBBCA-4017-5G COBBCA-4017-100G
4-Bromo-2-nitrobenzoic acid
4-Bromo-2-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MDL Number: MFCD01013599
CAS Number: 99277-71-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR