4-Bromo-2-fluorobenzaldehyde

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OS-7709-25G OS-7709-100G
COBBOS-7709-25GEA 0 GBP
COBBOS-7709-25G COBBOS-7709-100G
4-Bromo-2-fluorobenzaldehyde
4-Bromo-2-fluorobenzaldehyde
Formula: C₇H₄BrFO
MDL Number: MFCD00143261
CAS Number: 57848-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR