4-Bromo-2-chloro-5-methoxypyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7273-1G
PY-7273-250M
PY-7273-5G
COBBPY-7273-1GEA
0
GBP
COBBPY-7273-1G
COBBPY-7273-250M
COBBPY-7273-5G
4-Bromo-2-chloro-5-methoxypyridine
4-Bromo-2-chloro-5-methoxypyridine
Formula:
C₆H₅BrClNO |
MDL Number:
MFCD09972186 CAS Number: 1020253-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...