4-Aminobenzamidine dihydrochloride

Supplier: BACHEM BIOCHEMICA
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

4031123.0005 4031123.0025
BACE4031123.0005EA 0 GBP
BACE4031123.0005 BACE4031123.0025
4-Aminobenzamidine dihydrochloride
4-Aminobenzamidine dihydrochloride
Formula: C₇H₁₁Cl₂N₃
MDL Number: MFCD00013001
CAS Number: 2498-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR