4-Amino-2-methylpyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7356-1G PY-7356-5G
COBBPY-7356-1GEA 0 GBP
COBBPY-7356-1G COBBPY-7356-5G
4-Amino-2-methylpyridine
4-Amino-2-methylpyridine
Formula: C₆H₈N₂
MDL Number: MFCD00186509
CAS Number: 18437-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR