4-Amino-2-chloropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

BK-4501-25G BK-4501-100G
COBBBK-4501-25GEA 0 GBP
COBBBK-4501-25G COBBBK-4501-100G
4-Amino-2-chloropyridine
4-Amino-2-chloropyridine
Formula: C₅H₅ClN₂
Boiling Pt: 300 °C (1013 hPa)
Melting Pt: 87…89 °C
MDL Number: MFCD00060089
CAS Number: 14432-12-3
EINECS: 238-403-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR