4,5-Diiodo-1-methylimidazole

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4,5-Diiodo-1-methyl-1H-imidazole

HI-1027-5G HI-1027-25G HI-1027-1G
COBBHI-1027-5GEA 0 GBP
COBBHI-1027-5G COBBHI-1027-25G COBBHI-1027-1G
4,5-Diiodo-1-methylimidazole
4,5-Diiodo-1-methylimidazole
Formula: C₄H₄I₂N₂
MW: 333.9 g/mol
MDL Number: MFCD03001336
CAS Number: 37067-96-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR