Ethyl-4,4,4-trifluoroacetoacetate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001234-25G 001234-100G 001234-500G
FLUO001234-25GEA 0 GBP
FLUO001234-25G FLUO001234-100G FLUO001234-500G
Ethyl-4,4,4-trifluoroacetoacetate
Ethyl 4,4,4-trifluoroacetoacetate
Formula: C₆H₇F₃O₃
Boiling Pt: 131 °C (1013 hPa)
Melting Pt: –39 °C
Density: 1.2586 g/cm³ (15.5 °C)
MDL Number: MFCD00000424
CAS Number: 372-31-6
EINECS: 206-750-7
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR