4-(Trifluoromethyl)thiobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019513-1G 019513-25G 019513-5G
FLUO019513-1GEA 0 GBP
FLUO019513-1G FLUO019513-25G FLUO019513-5G
4-(Trifluoromethyl)thiobenzamide
4-(Trifluoromethyl)thiobenzamide
Formula: C₈H₆F₃NS
Boiling Pt: 254 °C (1013 hPa)
Melting Pt: 135 °C
MDL Number: MFCD00051806
CAS Number: 72505-21-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR