4-(Trifluoromethoxy)benzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OS-7494-100G OS-7494-25G OS-7494-5G
COBBOS-7494-100GEA 0 GBP
COBBOS-7494-100G COBBOS-7494-25G COBBOS-7494-5G
4-(Trifluoromethoxy)benzoic acid
4-(Trifluoromethoxy)benzoic acid
Formula: C₈H₅F₃O₃
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 153,4 °C
MDL Number: MFCD00002541
CAS Number: 330-12-1
EINECS: 206-352-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR