Methyl-4-(dimethylamino)benzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4655-5G CA-4655-25G
COBBCA-4655-5GEA 0 GBP
COBBCA-4655-5G COBBCA-4655-25G
Methyl-4-(dimethylamino)benzoate
Methyl 4-(dimethylamino)benzoate
Formula: C₁₀H₁₃NO₂
MDL Number: MFCD00014905
CAS Number: 1202-25-1
EINECS: 214-863-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR