4-(Difluoromethoxy)benzaldehyde
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7712-5G
OS-7712-25G
OS-7712-1G
COBBOS-7712-5GEA
0
GBP
COBBOS-7712-5G
COBBOS-7712-25G
COBBOS-7712-1G
4-(Difluoromethoxy)benzaldehyde
4-(Difluoromethoxy)benzaldehyde
Formula:
C₈H₆F₂O₂ MW: 172.13 g/mol |
MDL Number:
MFCD00042252 CAS Number: 73960-07-3 EINECS: 277-653-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...