3-Methylthiobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3-Methylbenzothioamide, Thio-m-toluamide

021340-1G 021340-5G
FLUO021340-1GEA 0 GBP
FLUO021340-1G FLUO021340-5G
3-Methylthiobenzamide
3-Methylthiobenzamide
Formula: C₈H₉NS
MDL Number: MFCD01314035
CAS Number: 2362-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR