Furan-3-boronic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

011038-5G 011038-1G
FLUO011038-5GEA 0 GBP
FLUO011038-5G FLUO011038-1G
Furan-3-boronic acid
Furan-3-boronic acid
Formula: C₄H₅BO₃
MDL Number: MFCD01319007
CAS Number: 55552-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR