3-Fluoro-5-methylpyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0076-5G OT-0076-25G OT-0076-1G
COBBOT-0076-5GEA 0 GBP
COBBOT-0076-5G COBBOT-0076-25G COBBOT-0076-1G
3-Fluoro-5-methylpyridine
3-Fluoro-5-methylpyridine
Formula: C₆H₆FN
MDL Number: MFCD03095273
CAS Number: 407-21-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR