3-Fluoro-4-(trifluoromethyl)pyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010727-1G 010727-2.5G
FLUO010727-1GEA 0 GBP
FLUO010727-1G FLUO010727-2.5G
3-Fluoro-4-(trifluoromethyl)pyridine
3-Fluoro-4-(trifluoromethyl)pyridine
Formula: C₆H₃F₄N
MDL Number: MFCD03789250
CAS Number: 113770-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR